Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 136115703-136115968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR4
Entrez Gene ID
7852
Gene Name
C-X-C motif chemokine receptor 4
Gene Aliases
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS
UniGene
Hs.593413
Gene Chromosome Location
Chr.2: 136114349-136118155 on Build GRCh38
Transcripts
NM_001008540.1 NM_003467.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4303872 c.56G>T p.G19V Chr.2: 136115872-136115872 on Build GRCh38
6975935 c.89G>T p.R30L Chr.2: 136115839-136115839 on Build GRCh38
4875746 c.76G>A p.E26K Chr.2: 136115852-136115852 on Build GRCh38
5465086 c.19C>A p.L7I Chr.2: 136115921-136115921 on Build GRCh38
3990611 c.129C>A p.I43I Chr.2: 136115811-136115811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768296315 Chr.2: 136115841-136115841 on Build GRCh38
rs533290861 Chr.2: 136115757-136115757 on Build GRCh38
rs770327175 Chr.2: 136115878-136115878 on Build GRCh38
rs147467366 Chr.2: 136115791-136115791 on Build GRCh38
rs757881953 Chr.2: 136115819-136115819 on Build GRCh38
rs774440989 Chr.2: 136115915-136115915 on Build GRCh38
rs779590491 Chr.2: 136115825-136115825 on Build GRCh38
rs748493255 Chr.2: 136115861-136115861 on Build GRCh38
rs781598359 Chr.2: 136115859-136115859 on Build GRCh38
rs1053292586 Chr.2: 136115926-136115926 on Build GRCh38
rs923351521 Chr.2: 136115839-136115839 on Build GRCh38
rs145879963 Chr.2: 136115775-136115775 on Build GRCh38
rs773216142 Chr.2: 136115740-136115740 on Build GRCh38
rs778969048 Chr.2: 136115800-136115800 on Build GRCh38
rs56400844 Chr.2: 136115771-136115771 on Build GRCh38
rs776615103 Chr.2: 136115936-136115936 on Build GRCh38
rs184978014 Chr.2: 136115829-136115829 on Build GRCh38
rs749938835 Chr.2: 136115818-136115818 on Build GRCh38
rs751756723 Chr.2: 136115762-136115762 on Build GRCh38
rs777105261 Chr.2: 136115878-136115878 on Build GRCh38
rs375868851 Chr.2: 136115889-136115889 on Build GRCh38
rs759904341 Chr.2: 136115929-136115929 on Build GRCh38
rs770275656 Chr.2: 136115730-136115730 on Build GRCh38
rs770986091 Chr.2: 136115908-136115908 on Build GRCh38
rs763059810 Chr.2: 136115750-136115750 on Build GRCh38
rs560844176 Chr.2: 136115909-136115909 on Build GRCh38
rs762966305 Chr.2: 136115891-136115891 on Build GRCh38
rs767838620 Chr.2: 136115930-136115930 on Build GRCh38
rs753680989 Chr.2: 136115780-136115780 on Build GRCh38

View Full Product Details