Gene Transcripts

Gene Symbol
Gatad2b
Entrez Gene ID
229542
Gene Name
GATA zinc finger domain containing 2B
Gene Aliases
AL118180, C430014D17Rik, P66beta, mKIAA1150
Chromosome Location
Chr. 3: 90293285 - 90363406 on Build GRCm38
UniGene ID
Mm.270999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139304.1 NP_647465.1 1 196
GenBank mRNA AF411837.1 - 1 196
AK041594.1 - 2 253
AK141586.1 - 3 483
BC130262.1 - 1 198 40142858

View Full Product Details