Gene Transcripts
- Gene Symbol
- Olfm3
- Entrez Gene ID
- 229759
- Gene Name
- olfactomedin 3
- Gene Aliases
- B230206G02Rik
- Chromosome Location
- Chr. 3: 114904078 - 115125764 on Build GRCm38
- UniGene ID
- Mm.54183
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286750.1 |
NP_001273679.1
|
2, 3 |
227 |
|
NM_153157.3 |
NP_694797.1
|
2, 3 |
378 |
|
NM_153458.3 |
NP_703188.1
|
2, 3 |
631 |
|
GenBank mRNA |
AF442824.1 |
- |
2, 3 |
378 |
|
AF442825.1 |
- |
2, 3 |
346 |
|
AK038569.1 |
- |
2, 3 |
578 |
|
AK044325.1 |
- |
2, 3 |
433 |
|
AK044605.1 |
- |
2, 3 |
581 |
|
AK045482.1 |
- |
2, 3 |
580 |
|
AK049612.1 |
- |
2, 3 |
632 |
|
AK079315.1 |
- |
2, 3 |
441 |
|
AK137414.1 |
- |
2, 3 |
433 |
|
BB628879.1 |
- |
2, 3 |
228 |
|
BC112422.1 |
- |
2, 3 |
287 |
40061966 |
BC113129.1 |
- |
2, 3 |
287 |
40061967 |
View Full Product Details