Gene Transcripts

Gene Symbol
Olfr816
Entrez Gene ID
258667
Gene Name
olfactory receptor 816
Gene Aliases
MOR113-1
Chromosome Location
Chr. 10: 129911338 - 129912276 on Build GRCm38
UniGene ID
Mm.377629
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146672.1 NP_666883.1 1 491
GenBank mRNA BC100682.1 - 1 489

View Full Product Details