Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175141.5 | NP_780350.1 | 5, 6 | 799 | |
GenBank mRNA | AK007661.1 | - | 5, 6 | 827 | |
AK078166.1 | - | 5, 6 | 800 | ||
AK146041.1 | - | 5, 6 | 821 | ||
AK169085.1 | - | 5, 6 | 806 | ||
BC038642.1 | - | 5, 6 | 787 | ||
BC051146.1 | - | 5, 6 | 811 | ||
BC137660.1 | - | 5, 6 | 892 | 8860679 |