Gene Transcripts
- Gene Symbol
- Slc25a12
- Entrez Gene ID
- 78830
- Gene Name
- solute carrier family 25 (mitochondrial carrier, Aralar), member 12
- Gene Aliases
- 2610002D09Rik, AI839531, B230107K20Rik, BB129864
- Chromosome Location
- Chr. 2: 71274270 - 71376694 on Build GRCm38
- UniGene ID
- Mm.146696
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172436.3 |
NP_766024.1
|
1, 2 |
43 |
|
GenBank mRNA |
AK019150.1 |
- |
1, 2 |
44 |
|
AK030595.1 |
- |
1, 2 |
73 |
|
AK083156.1 |
- |
1, 2 |
44 |
|
AK144958.1 |
- |
1, 2 |
77 |
|
AK147948.1 |
- |
1, 2 |
58 |
|
AK208043.1 |
- |
1, 2 |
110 |
|
BC013845.1 |
- |
1, 2 |
22 |
|
BC024952.1 |
- |
1, 2 |
238 |
|
BC060505.1 |
- |
1, 2 |
30 |
6311836 |
View Full Product Details