Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016669.1 | NP_057878.1 | 6, 7 | 866 | |
GenBank mRNA | AF039391.1 | - | 6, 7 | 866 | |
AK138422.1 | - | 6, 7 | 837 | ||
AK143116.1 | - | 6, 7 | 856 | ||
AK163781.1 | - | 6, 7 | 837 | ||
BC045159.1 | - | 6, 7 | 827 | 6492374 |