Gene Transcripts

Gene Symbol
Olfr822
Entrez Gene ID
258666
Gene Name
olfactory receptor 822
Gene Aliases
MOR113-2
Chromosome Location
Chr. 10: 130074412 - 130075350 on Build GRCm38
UniGene ID
Mm.377628
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146671.1 NP_666882.1 1 285
GenBank mRNA BC127972.1 - 1 289

View Full Product Details