Gene Transcripts

Gene Symbol
Tmem143
Entrez Gene ID
70209
Gene Name
transmembrane protein 143
Gene Aliases
2310076O21Rik, AI481604
Chromosome Location
Chr. 7: 45896990 - 45917418 on Build GRCm38
UniGene ID
Mm.163766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144801.2 NP_659050.2 5 663
GenBank mRNA AK039173.1 - 4 733
AK087636.1 - 4 868
BC017618.1 - 5 657 4923610

View Full Product Details