Gene Transcripts

Gene Symbol
Cgref1
Entrez Gene ID
245918
Gene Name
cell growth regulator with EF hand domain 1
Gene Aliases
Cgr11
Chromosome Location
Chr. 6: 26797126 - 26809132 on Build Rnor_6.0
UniGene ID
Rn.31842
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139087.2 NP_620787.2 6 956
GenBank mRNA BI279285.1 - 1 390
FQ220335.1 - 6 886
U66470.1 - 6 877

View Full Product Details