Gene Transcripts

Gene Symbol
Spta1
Entrez Gene ID
289257
Gene Name
spectrin, alpha, erythrocytic 1
Gene Aliases
Spna1
Chromosome Location
Chr. 13: 92264231 - 92340091 on Build Rnor_6.0
UniGene ID
Rn.44157
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011908.3 NP_001011908.2 16 2343
GenBank mRNA AY238342.1 - 16 2343

View Full Product Details