Gene Transcripts

Gene Symbol
Nme8
Entrez Gene ID
364729
Gene Name
NME/NM23 family member 8
Gene Aliases
Sptrx2, Txndc3
Chromosome Location
Chr. 17: 47301463 - 47368543 on Build Rnor_6.0
UniGene ID
Rn.107066
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198792.2 NP_942087.2 15 1615
XM_006254088.2 XP_006254150.1 15 1664
GenBank mRNA AF548544.1 - 13 1322
BC097265.1 - 14 1389

View Full Product Details