Gene Transcripts

Gene Symbol
Naaladl1
Entrez Gene ID
83568
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
Gene Aliases
Naaladasel
Chromosome Location
Chr. 1: 221470675 - 221484222 on Build Rnor_6.0
UniGene ID
Rn.10856
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031759.1 NP_113947.1 17 2063
GenBank mRNA AF009921.1 - 17 2063

View Full Product Details