Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_004119.2 | NP_004110.2 |
NR_130706.1 | ||
XM_011535015.2 | XP_011533317.1 | |
XM_011535017.2 | XP_011533319.1 | |
XM_011535018.2 | XP_011533320.1 | |
XM_017020486.1 | XP_016875975.1 | |
XM_017020487.1 | XP_016875976.1 | |
XM_017020488.1 | XP_016875977.1 | |
XM_017020489.1 | XP_016875978.1 | |
GenBank mRNA | AW057705.1 | - |
AW772610.1 | - | |
BC036028.1 | - | |
BC126350.1 | - | |
BC144039.1 | - | |
BC144040.1 | - | |
BI461248.1 | - | |
ENST00000241453.0 | - | |
L36162.1 | - | |
U02687.1 | - | |
Z26652.1 | - |