Gene Transcripts

Gene Symbol
Prl
Entrez Gene ID
19109
Gene Name
prolactin
Gene Aliases
AV290867, Prl1a1
Chromosome Location
Chr.13: 27057570 - 27065204 on Build GRCm38
Uni Gene
Mm.1270
Interrogated Sequence Translated Protein
RefSeq NM_001163530.1 NP_001157002.1
NM_011164.2 NP_035294.2
GenBank mRNA AK017520.1 -
AK017521.1 -
AK017547.1 -
AK017562.1 -
AK017579.1 -
AK017584.1 -
AK077493.1 -
AK133645.1 -
AK133736.1 -
AK133752.1 -
AK161553.1 -
AK161557.1 -
AK161565.1 -
AK161594.1 -
AK161619.1 -
BC061141.1 -
X02892.1 -
X04418.1 -

View Full Product Details