Gene Transcripts

Gene Symbol
Flt3
Entrez Gene ID
14255
Gene Name
FMS-like tyrosine kinase 3
Gene Aliases
B230315G04, CD135, Flk-2, Flk2, Flt-3, Ly72, wmfl
Chromosome Location
Chr.5: 147330741 - 147400489 on Build GRCm38
Uni Gene
Mm.194
Interrogated Sequence Translated Protein
RefSeq NM_010229.2 NP_034359.2
GenBank mRNA AK045865.1 -
AK149292.1 -
AK163640.1 -
BC109003.1 -
BC109004.1 -
L36163.1 -
M64689.1 -
X59398.1 -

View Full Product Details