Gene Transcripts

Gene Symbol
Prmt6
Entrez Gene ID
99890
Gene Name
protein arginine N-methyltransferase 6
Gene Aliases
AW124876, BB233495, Hrmt1l6
Chromosome Location
Chr. 3: 110246104 - 110250999 on Build GRCm38
UniGene ID
Mm.36115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024139.2 2 2879
GenBank mRNA AK087551.1 - 2 2427
AK087608.1 - 2 2880
AK157132.1 - 3 2833
BC066221.1 - 2 2856 30436952

View Full Product Details