Gene Transcripts
- Gene Symbol
- 2010016I18Rik
- Entrez Gene ID
- 69206
- Gene Name
- RIKEN cDNA 2010016I18 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 106481986 - 106485913 on Build GRCm38
- UniGene ID
- Mm.14816
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB645704.1 |
- |
2 |
323 |
|
Chr. 3 - 106482910 - 106485913 [-] |
GenBankmRNA |
BB645667.1 |
- |
2 |
323 |
|
Chr. 3 - 106482898 - 106485913 [-] |
GenBankmRNA |
BB215580.2 |
- |
1 |
361 |
|
Chr. 3 - 106482003 - 106483578 [-] |
View Full Product Details