Gene Transcripts
- Gene Symbol
- MGC15885
- Entrez Gene ID
- 197003
- Gene Name
- uncharacterized protein MGC15885
- Gene Aliases
- Chromosome Location
- Chr. 15: 62637172 - 62645181 on Build GRCh38
- UniGene ID
- Hs.352253
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026897.1 |
|
3 |
1213 |
|
GenBank mRNA |
BC014404.1 |
- |
3 |
1335 |
|
BC131619.1 |
- |
3 |
1112 |
|
View Full Product Details