Gene Transcripts
- Gene Symbol
- FAM222A-AS1
- Entrez Gene ID
- 84983
- Gene Name
- FAM222A antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 12: 109734209 - 109773487 on Build GRCh38
- UniGene ID
- Hs.675961
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ081961.1 |
- |
3 |
453 |
|
Chr. 12 - 109734425 - 109773444 [-] |
GenBankmRNA |
BF685112.1 |
- |
|
489 |
|
Chr. - - [-] |
GenBankmRNA |
DA559753.1 |
- |
2 |
441 |
|
Chr. 12 - 109734412 - 109735957 [-] |
GenBankmRNA |
BI222945.1 |
- |
4 |
503 |
|
Chr. 12 - 109734331 - 109773429 [-] |
GENCODE |
ENST00000541723.5 |
- |
3 |
517 |
|
Chr. 12 - 109734166 - 109773508 [-] |
View Full Product Details