Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199633.1 | NP_001186562.1 | 18 | 4376 | |
NM_022127.2 | NP_071410.1 | 19 | 4322 | ||
NR_037638.2 | 19 | 4552 | |||
XM_011518905.2 | XP_011517207.1 | 18 | 4446 | ||
XM_011518906.2 | XP_011517208.1 | 19 | 4542 | ||
XM_011518907.2 | XP_011517209.1 | 17 | 4261 | ||
XM_011518908.2 | XP_011517210.1 | 13 | 4281 | ||
GenBank mRNA | AK022042.1 | - | 1 | 2118 |