Gene Transcripts

Gene Symbol
EDEM3
Entrez Gene ID
80267
Gene Name
ER degradation enhancing alpha-mannosidase like protein 3
Gene Aliases
C1orf22
Chromosome Location
Chr. 1: 184690491 - 184754907 on Build GRCh38
UniGene ID
Hs.523811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC070173.1 - 1 819

View Full Product Details