Gene Transcripts

Gene Symbol
C1orf74
Entrez Gene ID
148304
Gene Name
chromosome 1 open reading frame 74
Gene Aliases
URLC4
Chromosome Location
Chr. 1: 209779208 - 209784730 on Build GRCh38
UniGene ID
Hs.497642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152485.3 NP_689698.1 2 3149
GenBank mRNA AK022798.1 - 1 40

View Full Product Details