Gene Transcripts
- Gene Symbol
- WBP2NL
- Entrez Gene ID
- 164684
- Gene Name
- WBP2 N-terminal like
- Gene Aliases
- GRAMD7, PAWP
- Chromosome Location
- Chr. 22: 41998725 - 42058456 on Build GRCh38
- UniGene ID
- Hs.655541
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038789.1 |
- |
4 |
664 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI461473.1 |
- |
4 |
669 |
|
Chr. 22 - 41998793 - 42002910 [+] |
View Full Product Details