Gene Transcripts
- Gene Symbol
- Gm53
- Entrez Gene ID
- 193022
- Gene Name
- predicted gene 53
- Gene Aliases
- Chromosome Location
- Chr. 11: 96251660 - 96264484 on Build GRCm38
- UniGene ID
- Mm.169508
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078566.1 |
- |
2 |
263 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB623715.1 |
- |
2 |
260 |
|
Chr. 11 - 96251771 - 96262232 [+] |
View Full Product Details