Gene Transcripts
- Gene Symbol
- 2010106G04Rik
- Entrez Gene ID
- 76533
- Gene Name
- RIKEN cDNA 2010106G04 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008322.1 |
- |
1 |
149 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708250.1 |
- |
1 |
146 |
|
Chr. 17 - 29614465 - 29614769 [-] |
View Full Product Details