Gene Transcripts
- Gene Symbol
- LOC105378137
- Entrez Gene ID
- 105378137
- Gene Name
- uncharacterized LOC105378137
- Gene Aliases
- Chromosome Location
- Chr. 6: 168225279 - 168226517 on Build GRCh38
- UniGene ID
- Hs.549668
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134590.1 |
|
2 |
259 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000424343.1 |
- |
2 |
259 |
|
Chr. 6 - 168225279 - 168226517 [-] |
View Full Product Details