Gene Transcripts
- Gene Symbol
- TUSC7
- Entrez Gene ID
- 285194
- Gene Name
- tumor suppressor candidate 7 (non-protein coding)
- Gene Aliases
- LINC00902, LSAMP-AS1, LSAMP-AS3, LSAMPAS3, NCRNA00295
- Chromosome Location
- Chr. 3: 116709788 - 116717040 on Build GRCh38
- UniGene ID
- Hs.120364
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000496242.5 |
- |
1 |
100 |
|
Chr. 3 - 116709969 - 116716431 [+] |
GENCODE |
ENST00000477539.6 |
- |
1 |
663 |
|
Chr. 3 - 116709406 - 116717321 [+] |
GENCODE |
ENST00000466156.1 |
- |
1 |
100 |
|
Chr. 3 - 116709969 - 116716431 [+] |
View Full Product Details