Gene Transcripts
- Gene Symbol
- LOC101929628
- Entrez Gene ID
- 101929628
- Gene Name
- uncharacterized LOC101929628
- Gene Aliases
- Chromosome Location
- Chr. 8: 61785072 - 62100736 on Build GRCh38
- UniGene ID
- Hs.116324
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL137390.1 |
- |
1 |
766 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000519990.1 |
- |
1 |
766 |
|
Chr. 8 - 61878882 - 61885562 [+] |
View Full Product Details