Gene Transcripts
- Gene Symbol
- LINC00540
- Entrez Gene ID
- 100506622
- Gene Name
- long intergenic non-protein coding RNA 540
- Gene Aliases
- Chromosome Location
- Chr. 13: 22210285 - 22276521 on Build GRCh38
- UniGene ID
- Hs.255773
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103810.1 |
|
2 |
472 |
|
GenBank mRNA |
AK054845.1 |
- |
2 |
739 |
|
BC035104.2 |
- |
2 |
472 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000616974.1 |
- |
|
472 |
|
Chr. - - [-] |
GENCODE |
ENST00000631321.1 |
- |
|
738 |
|
Chr. - - [-] |
GenBankmRNA |
BI548513.1 |
- |
2 |
476 |
|
Chr. 13 - 22210285 - 22275078 [+] |
View Full Product Details