Gene Transcripts
- Gene Symbol
- Gm11770
- Entrez Gene ID
- 105243647
- Gene Name
- predicted gene 11770
- Gene Aliases
- OTTMUSG00000004111
- Chromosome Location
- Chr. 11: 120213034 - 120222100 on Build GRCm38
- UniGene ID
- Mm.406381
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084828.1 |
- |
2 |
491 |
|
View Full Product Details