Gene Transcripts
- Gene Symbol
- Gm13157
- Entrez Gene ID
- 100041677
- Gene Name
- predicted gene 13157
- Gene Aliases
- OTTMUSG00000010679
- Chromosome Location
- Chr. 4: 147753974 - 147810843 on Build GRCm38
- UniGene ID
- Mm.327194
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK163132.1 |
- |
4 |
575 |
|
View Full Product Details