Gene Transcripts

Gene Symbol
Trim59
Entrez Gene ID
66949
Gene Name
tripartite motif-containing 59
Gene Aliases
2310035M22Rik, 2700022F13Rik, Mrf1, TSBF1
Chromosome Location
Chr. 3: 69035292 - 69046404 on Build GRCm38
UniGene ID
Mm.176695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025863.3 NP_080139.3 3 422
GenBank mRNA AK012269.1 - 3 423
BC006700.1 - 3 340 3501285
BC025430.1 - 3 355 4017983

View Full Product Details