Gene Transcripts

Gene Symbol
Atp2a3
Entrez Gene ID
25391
Gene Name
ATPase, Ca++ transporting, ubiquitous
Gene Aliases
SERCA3
Chromosome Location
Chr. 10: 59528824 - 59560440 on Build Rnor_6.0
UniGene ID
Rn.9920
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012914.1 NP_037046.1 16 2520
XM_006246597.2 XP_006246659.1 17 3134
GenBank mRNA M30581.1 - 16 2520

View Full Product Details