Gene Transcripts

Gene Symbol
Sh3gl1
Entrez Gene ID
81922
Gene Name
SH3-domain GRB2-like 1
Gene Aliases
SH3P8
Chromosome Location
Chr. 9: 11031847 - 11055285 on Build Rnor_6.0
UniGene ID
Rn.8762
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031239.2 NP_112518.1 3 310
GenBank mRNA AB008161.1 - 3 310
AF009602.1 - 3 295
BC070893.1 - 3 306 7099223

View Full Product Details