Gene Transcripts

Gene Symbol
Grifin
Entrez Gene ID
117130
Gene Name
galectin-related inter-fiber protein
Gene Aliases
Chromosome Location
Chr. 12: 16170108 - 16172144 on Build Rnor_6.0
UniGene ID
Rn.26894
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057187.2 NP_476535.1 3, 4 265
XM_008768938.1 XP_008767160.1 3, 4 319
GenBank mRNA AF082160.1 - 3, 4 265

View Full Product Details