Gene Transcripts

Gene Symbol
Tmem9
Entrez Gene ID
289046
Gene Name
transmembrane protein 9
Gene Aliases
Chromosome Location
Chr. 13: 52854031 - 52871300 on Build Rnor_6.0
UniGene ID
Rn.2747
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105953.2 NP_001099423.2 4 308
NM_001277263.1 NP_001264192.1 4 385
GenBank mRNA BC169086.1 - 3 275

View Full Product Details