Gene Transcripts

Gene Symbol
Hmgn3
Entrez Gene ID
94353
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
1110002A15Rik, 6330514M13Rik, BB071015, TRIP7
Chromosome Location
Chr. 9: 83109942 - 83146734 on Build GRCm38
UniGene ID
Mm.244426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026122.4 NP_080398.1 5 334
NM_175074.2 NP_778249.1 5 334
GenBank mRNA AK002970.1 - 5 253
AK003272.1 - 5 282
AK153223.1 - 5 294
BC005693.1 - 5 270 3597594
CJ150650.1 - 5 331

View Full Product Details