Gene Transcripts

Gene Symbol
C19orf84
Entrez Gene ID
147646
Gene Name
chromosome 19 open reading frame 84
Gene Aliases
Chromosome Location
Chr. 19: 51388289 - 51390574 on Build GRCh38
UniGene ID
Hs.148344
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193623.1 NP_001180552.1 2 1026
NR_126530.1 2 1155
GenBank mRNA BC043532.1 - 2 1026
BC085012.1 - 2 999

View Full Product Details