Gene Transcripts
- Gene Symbol
- Ifna15
- Entrez Gene ID
- 242517
- Gene Name
- interferon alpha 15
- Gene Aliases
- Gm12597, Ifnaa, MuIFN-alpha-A, OTTMUSG00000007655
- Chromosome Location
- Chr. 4: 88557673 - 88558245 on Build GRCm38
- UniGene ID
- Mm.377261
- Species
- Mus musculus
- Gene Symbol
- Ifna12
- Entrez Gene ID
- 242519
- Gene Name
- interferon alpha 12
- Gene Aliases
- Chromosome Location
- Chr. 4: 88602580 - 88603376 on Build GRCm38
- UniGene ID
- Mm.246618
- Species
- Mus musculus
- Gene Symbol
- Ifna13
- Entrez Gene ID
- 230396
- Gene Name
- interferon alpha 13
- Gene Aliases
- Chromosome Location
- Chr. 4: 88643641 - 88644459 on Build GRCm38
- UniGene ID
- Mm.246592
- Species
- Mus musculus
- Gene Symbol
- Ifna14
- Entrez Gene ID
- 404549
- Gene Name
- interferon alpha 14
- Gene Aliases
- OTTMUSG00000007662
- Chromosome Location
- Chr. 4: 88571229 - 88571798 on Build GRCm38
- UniGene ID
- Mm.377947
- Species
- Mus musculus
- Gene Symbol
- Ifna6
- Entrez Gene ID
- 15969
- Gene Name
- interferon alpha 6
- Gene Aliases
- IFNa8/6, Ifa6, Ifa8, Ifna8
- Chromosome Location
- Chr. 4: 88827416 - 88827985 on Build GRCm38
- UniGene ID
- Mm.377090
- Species
- Mus musculus
- Gene Symbol
- Ifnab
- Entrez Gene ID
- 15974
- Gene Name
- interferon alpha B
- Gene Aliases
- Chromosome Location
- Chr. 4: 88690599 - 88691268 on Build GRCm38
- UniGene ID
- Mm.377093
- Species
- Mus musculus
View Full Product Details