Gene Transcripts

Gene Symbol
Pde6h
Entrez Gene ID
114248
Gene Name
phosphodiesterase 6H, cGMP-specific, cone, gamma
Gene Aliases
Pde6g
Chromosome Location
Chr. 4: 170946694 - 170963327 on Build Rnor_6.0
UniGene ID
Rn.21249
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053688.2 NP_446140.1 1 103
XM_006237558.2 XP_006237620.1 2 1431
GenBank mRNA AF169390.1 - 1 93

View Full Product Details