Gene Transcripts

Gene Symbol
HES2
Entrez Gene ID
54626
Gene Name
hes family bHLH transcription factor 2
Gene Aliases
bHLHb40
Chromosome Location
Chr. 1: 6415232 - 6419919 on Build GRCh38
UniGene ID
Hs.118727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019089.4 NP_061962.2 4 1556
GenBank mRNA AK023754.1 - 1 740
AK091122.1 - 4 1556

View Full Product Details