Gene Transcripts

Gene Symbol
CACNG6
Entrez Gene ID
59285
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Gene Aliases
Chromosome Location
Chr. 19: 53991149 - 54012666 on Build GRCh38
UniGene ID
Hs.631560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031897.2 NP_114103.2 2 1515
NM_145814.1 NP_665813.1 4 1728
NM_145815.1 NP_665814.1 3 1590
NR_102308.1 3 718
GenBank mRNA BC021552.1 - 4 1728

View Full Product Details