Gene Transcripts

Gene Symbol
Ap3d1
Entrez Gene ID
11776
Gene Name
adaptor-related protein complex 3, delta 1 subunit
Gene Aliases
AA407035, Ap3d, Bolvr, mBLVR1, mh, mocha
Chromosome Location
Chr. 10: 80706956 - 80742282 on Build GRCm38
UniGene ID
Mm.209294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007460.1 NP_031486.1 31 3743
GenBank mRNA AB004305.1 - 31 3743
BC048786.1 - 31 3733 6489789
BC053066.1 - 31 3759 5704929

View Full Product Details