Gene Transcripts
- Gene Symbol
- Nfkb2
- Entrez Gene ID
- 18034
- Gene Name
- nuclear factor of kappa light polypeptide gene enhancer in B cells 2, p49/p100
- Gene Aliases
- NF-kappaB2, lyt, p49, p49/p100, p50B, p52
- Chromosome Location
- Chr. 19: 46303612 - 46312400 on Build GRCm38
- UniGene ID
- Mm.102365
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177369.1 |
NP_001170840.1
|
6, 7 |
552 |
|
NM_001177370.1 |
NP_001170841.1
|
5, 6 |
468 |
|
NM_019408.3 |
NP_062281.1
|
5, 6 |
703 |
|
GenBank mRNA |
AF155372.1 |
- |
6, 7 |
630 |
|
AF155373.1 |
- |
6, 7 |
667 |
|
AK134081.1 |
- |
6, 7 |
971 |
|
AK137680.1 |
- |
6, 7 |
542 |
|
AK144994.1 |
- |
6, 7 |
545 |
|
AK145312.1 |
- |
5, 6 |
669 |
|
AK148160.1 |
- |
6, 7 |
540 |
|
AK150012.1 |
- |
6, 7 |
531 |
|
AK153412.1 |
- |
1, 2 |
112 |
|
AK193270.1 |
- |
1, 2 |
132 |
|
AK216302.1 |
- |
1, 2 |
132 |
|
BC027423.1 |
- |
5, 6 |
469 |
3672913 |
DQ020182.1 |
- |
5, 6 |
400 |
|
DQ020183.1 |
- |
4, 5 |
521 |
|
DQ020184.1 |
- |
4, 5 |
521 |
|
DQ020185.1 |
- |
5, 6 |
400 |
|
View Full Product Details