Gene Transcripts
- Gene Symbol
- Rrs1
- Entrez Gene ID
- 59014
- Gene Name
- RRS1 ribosome biogenesis regulator homolog (S. cerevisiae)
- Gene Aliases
- 5730466A07Rik, AA415037, D1Ertd701e, Rrr
- Chromosome Location
- Chr. 1: 9545408 - 9547455 on Build GRCm38
- UniGene ID
- Mm.29061
- Species
- Mus musculus
- Gene Symbol
- Gm5067
- Entrez Gene ID
- 272714
- Gene Name
- predicted gene 5067
- Gene Aliases
- EG272714
- Chromosome Location
- Chr. 5: 26007007 - 26008551 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031269.1 |
- |
|
338 |
|
View Full Product Details