Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159369.1 | NP_001152841.1 | 11 | 2922 | |
NM_029977.2 | NP_084253.1 | 16 | 3759 | ||
GenBank mRNA | AK156001.1 | - | 7 | 2227 | |
AY074936.1 | - | 16 | 3759 | ||
AY147862.1 | - | 11 | 2922 | ||
AY147863.1 | - | 17 | 3855 | ||
AY147864.1 | - | 12 | 3018 | ||
BC138360.1 | - | 16 | 3700 | 8861380 | |
BC157900.1 | - | 16 | 3700 | 9088092 |