Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002238.3 | NP_002229.1 | 8 | 1688 | |
NM_172362.2 | NP_758872.1 | 8 | 1769 | ||
XM_017001246.1 | XP_016856735.1 | 3 | 907 | ||
GenBank mRNA | AF078741.1 | - | 8 | 1554 | |
AF078742.1 | - | 8 | 1635 | ||
AJ001366.1 | - | 8 | 1688 | ||
BC113709.1 | - | 8 | 1588 | 8322761 | |
BC143599.1 | - | 8 | 1589 | 9052111 |