Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1148 | |
GenBank mRNA | AJ409320.1 | - | 2 | 1007 | |
AK055556.1 | - | 2 | 1086 | ||
AK223574.1 | - | 2 | 1086 | ||
BC004299.1 | - | 2 | 1075 | ||
BC071947.1 | - | 2 | 1074 | 6459570 | |
BT006693.1 | - | 2 | 1007 |