Gene Transcripts

Gene Symbol
FAM210B
Entrez Gene ID
116151
Gene Name
family with sequence similarity 210 member B
Gene Aliases
5A3, C20orf108, dJ1167H4.1
Chromosome Location
Chr. 20: 56358927 - 56368663 on Build GRCh38
UniGene ID
Hs.143736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080821.2 NP_543011.2 3 584
GenBank mRNA AJ311123.1 - 3 545
AK075565.1 - 3 531
AK314768.1 - 3 569
BC017725.1 - 3 541 3887324

View Full Product Details